Learn More
Troemner™ Individual Analytical Precision Weights, Class 3
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
$157.38 - $3740.65
Specifications
| Classification | ASTM Class 3 |
|---|---|
| Includes | Polycarbonate case |
| Quantity | Each |
| Packaging Quantity | 1 |
| Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
|
02-214-029
|
Troemner™
80858669 |
2 mg |
Each for $187.26
|
|
|||||
|
02-215-25G
|
N/A | 100 g |
Each for $390.86
|
|
|||||
|
02214032
|
Troemner™
80858537 |
20 mg |
Each for $157.38
|
|
|||||
|
02-214-041
|
Troemner™
80858087 |
5 kg |
Each for $3,740.65
|
|
|||||
|
02-214-039
|
N/A | 1 kg |
Each for $1,828.41
|
|
|||||
|
02-215-25A
|
N/A | 1 g |
Each for $245.38
|
|
|||||
|
02-215-25D
|
N/A | 10 g |
Each for $321.34
|
|
|||||
|
02214036
|
Troemner™
80858289 |
2 g |
Each for $209.82
|
|
|||||
|
02-214-031
|
Troemner™
80858577 |
10 mg |
Each for $187.26
|
|
|||||
|
02-215-20H
|
N/A | 1 mg |
Each for $199.83
|
|
|||||
|
02-225-551
|
N/A | 500 mg |
Each for $200.76
|
|
|||||
|
02-214-033
|
Troemner™
80858486 |
50 mg |
Each for $188.06
|
|
|||||
|
02-214-038
|
N/A | 200 g |
Each for $570.62
|
|
|||||
|
02-214-037
|
Troemner™
80858275 |
3 g |
Each for $249.29
|
|
|||||
|
02214040
|
Troemner™
80858114 |
2 kg |
Each for $2,429.21
|
|
|||||
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1g to 0.01g
- Weights 1mg through 2kg are supplied in a high-quality polycarbonate case
- Reference standards as well as calibrating Class I balances as specified in OIML R 76
- Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Specifications
| ASTM Class 3 | |
| Polycarbonate case | |
| Each | |
| 1 |