missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-3-butyric acid, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210204205

 View more versions of this product

Catalog No. ICN10204205


Explore available promotions

Description

Description

Specifications

Specifications

Indole-3-butyric acid
133-32-4
C12H13NO2
5 g
JTEDVYBZBROSJT-UHFFFAOYSA-N
4-(1H-indol-3-yl)butanoic acid
8617
203.24
Plant growth hormone
White
MFCD00005664
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
203.241
CHEBI:33070
Powder

Chemical Identifiers

133-32-4
203.241
JTEDVYBZBROSJT-UHFFFAOYSA-N
8617
4-(1H-indol-3-yl)butanoic acid
C12H13NO2
MFCD00005664
indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
CHEBI:33070
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-101-5

Recommended Storage : Store at +4°C

SDS
Documents

Documents

Promotions

Promotions