missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-6-methanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 455542500
Specifications
| Indole-6-methanol | |
| Authentic | |
| C9H9NO | |
| 250mg | |
| WRMZOPANDOHWJU-UHFFFAOYSA-N | |
| 1H-indol-6-ylmethanol | |
| 2773459 | |
| 97% |
| 1075-26-9 | |
| 96% min. (GC) | |
| MFCD02179595 | |
| 6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol | |
| OCC1=CC=C2C=CNC2=C1 | |
| 147.18 | |
| 147.17 |
Chemical Identifiers
| 1075-26-9 | |
| 147.18 | |
| WRMZOPANDOHWJU-UHFFFAOYSA-N | |
| 2773459 | |
| OCC1=CC=C2C=CNC2=C1 |
| C9H9NO | |
| MFCD02179595 | |
| 6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol | |
| 1H-indol-6-ylmethanol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO â Research Use Only