missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Indomethacin

Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID).

Supplier:  Thermo Scientific Chemicals 458031000

 View more versions of this product

Catalog No. AC458031000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release

• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics

• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation

Applications

• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions

• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin

Specifications

Specifications

Indomethacin
155.0°C
Authentic
100 g
CGIGDMFJXJATDK-UHFFFAOYSA-N
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
3715
357.79
53-86-1
0.5% max. (100°C, 2 h, 5 mm Hg)
C19H16ClNO4
indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
357.79
CHEBI:49662
≥97.5% (HPLC)

Chemical Identifiers

53-86-1
357.79
indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
CHEBI:49662
CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
C19H16ClNO4
CGIGDMFJXJATDK-UHFFFAOYSA-N
3715
2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
May cause an allergic skin reaction.
May damage fertility.
May damage the unborn child.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use

GHS Signal Word: Danger

EINECSNumber : 200-186-5

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only