Fisherbrand™ Infecon™ 3000 Infectious Substance Shipper
Combines U.N. certification requirements with new innovative safety features
$1149.88 - $1149.88
Specifications
Applications For infectious substances
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
19-037-436
View Documents
Fisherbrand™
INF3000
Infectious Substance Shipper
Case of 12 Each for $1,149.88
Description
Description
  • Each shipper consists of a transparent pressure vessel able to withstand up to 95kPa, cushioning materials, outer box, labels, declarations and instructions
  • Transparent pressure vessel allows contents to be seen
  • Use the Infecon 3000 with the Speci-Freez™ Insulated Shipper (19-075-384C) for refrigerated or frozen specimens
  • Reusable when using the complete refurbishment kit (19-037-437)
Order Info
Dimensions: 6 x 6 x 8 in. (15.24 x 15.24 x 20.32cm) with 1L vessel
Specifications
Specifications
For infectious substances
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'