Mirus Bio™ Ingenio™ Cuvettes
Ingenio™ electroporation cuvettes for high efficiency electroporation using any instrument.
Supplier: Mirus Bio™ MIR50123
Description
- High Efficiency Electroporation– Deliver to hard to transfect cell lines and primary cells
- Compatible with Most Conventional Electroporation Devices – Use your existing system including Ingenio EZporator, Lonza-Amaxa, Bio-Rad or Harvard BTX
- Save Money and Reduce Research Costs Without Sacrificing Performance – Ingenio Electroporation
- Solution is available as a stand-alone solution or as part of a complete kit with cuvettes and cell droppers
- Developed the Ingenio Electroporation Solution to facilitate efficient and reliable delivery of nucleic acids to eukaryotic cells traditionally resistant to chemical transfection
- Ingenio is a broad spectrum solution that supports high efficiency electroporation with minimal toxicity.
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- Compatible with multiple instruments and facilitates a wide range of applications requiring nucleic acid delivery to cells
Specifications
Sterile | |
Reusable | |
Polycarbonate | |
50 Pk. | |
Electroporation |
Cuvette | |
800 μL | |
50 Cuvettes | |
EZporator Electroporation System |