missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mirus Bio™ Ingenio™ Electroporation Solution
Proprietary Ingenio™ Electroporation Solution facilitates efficient and reliable delivery of nucleic acids to eukaryotic cells resistant to chemical transfection.
Supplier: Mirus Bio™ MIR50111
Description
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications
Electroporation | |
Electroporation Solution | |
6.25 mL | |
4°C |
All electroporation device | |
25 Reactions | |
Sterile | |
Exponential Decay Wave |