missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mirus Bio™ Ingenio™ EZporator™ Electroporation System
Mirus Bio™ Ingenio™ EZporator™ Electroporation System provides a robust and universal approach for multiple insect and mammalian cell culture applications, including gene expression, RNAi, stable cell line generation, and CRISPR/Cas genome editing.
Supplier: Mirus Bio™ MIR51000
Description
- Digital Liquid Crystal Display (LCD) user interface
- Pulse Generator to deliver exponential decay pulse at user-determined voltage
- Cuvette chamber compatible with 0.1 cm, 0.2 cm or 0.4 cm cuvettes
- Power cords with Type B, F and G electrical outlet plugs
- Technical Specs: Low Voltage (LV) mode and High Voltage (HV) mode
- Dual monitoring and display of peak output voltage and time constant
- Streamlined workflow
- Initialize instrument
- Set voltage and mode with Voltage Adjust knob
- Deliver Electroporation Pulse button
- European regulations and directives: 2014/35/EU Low Voltage Directive (LVD), 2014/30/EU Electromagnetic Compatibility directive (EMC), 2012/19/EU Waste electrical and electronic equipment directive (WEEE), 2011/65/EU Restriction on the use of certain hazardous substances directive (RoHS)
- Standards to Demonstrate Conformity: EN 61010-1:2010 Safety requirements for electrical equipment for measurement,control and laboratory use, General Requirements EN 61326-1:2013 Electrical equipment for measurement, control and laboratory use
- EMC requirements: EN 50581:2012 Assessment of products with respect to RoHS
EMC and Safety testing performed by Intertek Testing Services - Listing Number 9801781
- Test reports: 103276746BOX-001, -002, -003
- Conforms to UL61010-1:2012
- Certified to CSA C22.2#61010-1:2012
Specifications
Mammalian and insect cell transfection | |
50/60 Hz | |
Electroporation System | |
30 to 2500 V with 10 V resolution | |
16-character Backlit LCD | |
Ezporator™ Pulse Generator (quantity 1)â, Ezporator™ Cuvette Chamber (quantity 1)â, Power cords (quantity 3 â Type B, F and G)â | |
22 cm | |
50°F to 104°F | |
10°C to 40°C | |
60% | |
25 cm | |
Non-sterile | |
1050 μF | |
5 sec. | |
Exponential Decay Wave |
Ingenio Electroporation Kits and Solution | |
Metal Case | |
20 to 2500 V | |
20 to 400 V with 2 V resolution | |
Exponential decay | |
22 cm | |
1 year warranty | |
5 ms to 125 ms | |
1 Ea. | |
25 cm | |
Voltage: 100 to 240 Vac, 50/60 Hz, CAT I; Power: 800 W; Fusion: 2.5 A, T rating 250 V | |
36 μF | |
Electrode Connector Cable | |
Digital LCD |