missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

METTLER TOLEDO™ InLab™ Reach pH Electrodes

A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.

Supplier:  METTLER TOLEDO™ 30248826

 View more versions of this product

Catalog No. 01-914-304


Explore available promotions

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pilot reactor
  • Tank and Barrel
  • Large samples
  • Deep vessels

Accessories & Care

  • Ceramic junction
  • HA glass membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Recommended Storage

InLab storage solution

Specifications

Specifications

Multi-Pin
Glass
Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ)
Multi-Pin to BNC (30281896)
HA Glass
InLab storage solution
225 mm
Ceramic
0 to 14
Accurate measurement in a wide range of sample types
3mol/L KCl
InLab™ Reach Pro-225
0°C to 100°C
12 mm
Videos
SDS
Documents

Documents

Promotions

Promotions