missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Reach pH Electrodes
A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.
Supplier: METTLER TOLEDO™ 30248828
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Pilot reactor
- Tank and Barrel
- Large samples
- Deep vessels
Accessories & Care
- Ceramic junction
- HA glass membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
pH Electrode InLab Reach Pt1000-225 | |
Ceramic | |
0 to 14 | |
Accurate measurement in a wide range of sample types | |
3mol/L KCl | |
InLab™ Reach Pt1000-225 | |
InLab storage solution | |
225 mm |
Multi-Pin | |
Glass | |
Combination Electrode; Integrated Temperature Probe (Pt-1000) | |
Multi-Pin to BNC (30281896) | |
HA Glass | |
Pt1000 | |
0°C to 100°C | |
12 mm |