Mettler Toledo™ InLab™ Sensor Cables: MultiPin™ Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter with compatible connector.
Supplier:  Mettler Toledo™ 30281897
 View more versions of this product
Catalog No. 01-914-266
Explore available promotions
Description
Description
  • Detachable
  • Resuable
Specifications
Specifications
MultiPin-BNC/RCA (cinch)
To connect Mettler Toledo InLab sensors to any meter
3.0 m
Mettler Toledo InLab Sensors
9.8 ft.