missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Surface pH Electrodes
A pH electrode featuring a flat, rugged plastic membrane for measuring samples with flat surfaces.
Supplier: METTLER TOLEDO™ 30249570
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Agar plates
- Drop size samples
- Paper
- Skin
- Leather
- Suspended solids
- Textiles and Prints
Accessories & Care
- Ceramic ring junction
- Polysulfone membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
Multi-Pin | |
Glass Shaft | |
Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology | |
Multi-Pin to BNC/RCA (30281896) | |
LoT Glass | |
InLab storage solution | |
120 mm |
Ceramic | |
1 to 11 | |
Measurement of solid or semi-solid samples | |
3mol/L KCl | |
InLab™ Surface Pro-ISM | |
0°C to 50°C | |
12 mm |