Innovive Card Holder
100% PET plastic cage card holder
Supplier:  Innovive CRDHLDU
Catalog No. 36-101-2381
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
For use with both mouse and rat disposable cages from Innovive.
Specifications
Specifications
PET
Mouse and rat disposable cages from Innovive
Up to ten cards