missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle
Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.
Supplier: Fisherbrand™ 10401133
Description
Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.
- Ideal for selecting, inoculating, or transferring microbiological samples
- Cools rapidly after sterilization
- Loops Slide along agar surfaces without cutting or tearing material
- Nichrome wire available in single or twisted configuration
- Passivated
- 203 mm (8 in.) handles, available with insulation
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Certifications
ISO 9001-02, GMP, cGMP
Specifications
11 in. | |
280 mm | |
Silver | |
Petri-Dish etc | |
Nichrome wire with insulated handle | |
Autoclavable |
Nichrome | |
3 mm | |
Inoculating Loop | |
25 | |
Available as Each or Pk. of 12 | |
Ideal for selecting, inoculating, or transferring microbiological samples |