missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Inositol, NF, 97-102%, Spectrum™ Chemical

I1012, 87-89-8, C6H12O6

Supplier:  Spectrum Chemical Mfg Cor I101212KG

 View more versions of this product

Catalog No. 18603455

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Spectrum™ Chemical Inositol, NF
Specifications

Specifications

87-89-8
Poly Pail
12 kg
OC1C(O)C(O)C(O)C(O)C1O
180.16
NF
100%
C6H12O6
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
97 to 102%

Chemical Identifiers

87-89-8
180.16
cyclohexane-1,2,3,4,5,6-hexol
C6H12O6
CDAISMWEOUEBRE-UHFFFAOYSA-N
OC1C(O)C(O)C(O)C(O)C1O
Videos
SDS
Documents

Documents

Promotions

Promotions