missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ Insert for 2 mL Standard Opening Vial

Allow the use of smaller sample volumes with standard autosampler vials

Supplier:  MilliporeSigma™ Supelco™ 27400U

 View more versions of this product

Catalog No. 111010956


Explore available promotions

Description

Description

  • Glass conical inserts with polymer feet are used to contain standard mixtures for various experimental purposes.
  • Vial inserts allow the use of smaller sample volumes with standard autosampler vials.
  • Inserts are generally chosen based on the inner diameter of the vial neck.
  • Standard opening vials (4.6 mm ID) require a 5 mm OD insert.
Specifications

Specifications

4.6 mm
Conical Clear Glass
100
Bottom Spring
4 mm
0.05 mL
25 mm
Videos
SDS
Documents

Documents

Promotions

Promotions