missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Insta-Chek™ Wide-Range Paper Dispenser

Single color match for each pH value.

$29.87 - $29.87

Specifications

pH Range 0 to 13
Includes Color chart
Quantity Each
View More Specs

Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
S60149
View Documents
Customer Channels Group (Ccg)
HJ613EMD
Paper Dispenser
Each for $29.87
Case of 10 Each for $298.70
 
Description

Description

  • 14 distinct colors match the 14 pH values for accurate readings
  • Includes color chart
  • Dispenser contains 50 ft. × 0.5 in.W roll (15.2m × 1.27cm)

Specifications

Specifications

0 to 13
Color chart
Each
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'