LaMotte™ Insta-TEST™ Low Range Phosphate Test Strips
Measures phosphate levels from 0, 100, 200, 300, 500, 1000, 2500 ppb in 15 seconds
Supplier: LaMotte™ 3021GENV
Description
- 25 strips per bottle
Specifications
Phosphate levels | |
Pool and Spa, Industrial Water Boiling and Cooling | |
0 to 2500 ppb | |
Phosphate Test Strips |
Strip | |
25 Strips/Bottle | |
15 sec. |