missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Insulin B (22-25)

Catalog No. AAJ61170MB
Change view
Click to view available options
Quantity:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61170MB 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AAJ61170MB missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' J61170MB
Only null left

For research use

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Formulation Lyophilized powder
Quantity 25mg
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length H-Arg-gly-Phe-Phe-OH
Product Type Insulin B (22-25)
Recombinant Native
Form Lyophilized

RUO – Research Use Only