missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Integra™ Miltex™ Instrument Tray and Cover
Constructed with 0.6mm stainless steel
$76.12 - $122.53
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
12-460-433
|
Integra™ Miltex™
MLT3949 |
Flat Cover |
Each for $76.12
|
|
|||||
12-460-432
|
Integra™ Miltex™
3948 |
Instrument Tray |
Each for $122.53
|
|
|||||
Specifications
Stainless Steel | |
0.6 mm |