Integra™ Miltex™ Scalpel Handles
Supplier:  Integra™ Miltex™ MLT410
 View more versions of this product
Catalog No. 12-460-143
Explore available promotions
Description
Description
  • Available in stainless steel and autoclavable plastic
  • Reusable
Specifications
Specifications
Stainless Steel
# 3L
21.6 cm
Reusable
Scalpel Handle
8.5 in.
# 10 to # 15C Blade