missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Integra™ Miltex™ Sterile Safety Scalpels
Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions
$63.17 - $63.17
Specifications
Blade Material | Stainless Steel |
---|---|
Product Type | Safety Scalpel |
Disposable | Single-use |
Sterility | Sterile |
Certifications/Compliance | OSHA |
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity | |||||
12-460-459
|
Integra™ Miltex™
MLT4511 |
# 11 | – |
Pack of 10 Each for $63.17
|
|
|||||
12-460-458
|
Integra™ Miltex™
MLT4510 |
# 10 | – |
Pack of 10 Each for $63.17
|
|
|||||
12-460-460
|
Integra™ Miltex™
MLT4515 |
# 15 | – |
Pack of 10 Each for $63.17
|
|
|||||
12-460-462
|
Integra™ Miltex™
MLT4522 |
# 22 | – |
Pack of 10 Each for $63.17
|
|
|||||
12-460-461
|
Integra™ Miltex™
4515C |
# 15 | With Angled Blade |
Pack of 10 Each for $63.17
|
|
|||||
Description
- Sterile
- Stainless-steel, razor-sharp cutting edge
- Metric ruler on each scalpel
- Individually packaged
Specifications
Stainless Steel | |
Single-use | |
OSHA |
Safety Scalpel | |
Sterile | |
Plastic |