missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Integra™ Miltex™ Sterile Standard Scalpels
Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions
$54.47 - $59.44
Specifications
Blade Material | Stainless Steel |
---|---|
For Use With (Application) | Superior Cutting and Clean Incisions |
Disposable | Single-use |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
12-460-451
|
Integra™ Miltex™
4410 |
# 10 |
Pack of 10 Each for $59.44
|
|
|||||
12-460-452
|
Integra™ Miltex™
4411 |
# 11 |
Pack of 10 Each for $59.44
|
|
|||||
12-460-454
|
Integra™ Miltex™
4420 |
# 20 |
Pack of 10 Each for $54.47
|
|
|||||
12-460-453
|
Integra™ Miltex™
MLT4415 |
# 15 |
Pack of 10 Each for $59.44
|
|
|||||
12-460-455
|
Integra™ Miltex™
MLT4421 |
# 21 |
Pack of 10 Each for $54.47
|
|
|||||
12-460-456
|
Integra™ Miltex™
4422 |
# 22 |
Pack of 10 Each for $59.44
|
|
|||||
12-460-457
|
Integra™ Miltex™
MLT4423 |
# 23 |
Pack of 10 Each for $54.47
|
|
|||||
Description
- Sterile
- Stainless steel, razor-sharp cutting edge
- Safety sleeve for blade
- Individually packaged
- Sold in Pack of 10
Specifications
Stainless Steel | |
Superior Cutting and Clean Incisions | |
Single-use | |
Sterile |