Integra™ Miltex™ Swiss Jeweler-Style Forceps
Supplier:  Integra™ MLT17302
 View more versions of this product
Catalog No. 12-460-242
Explore available promotions
Description
Description
  • Stainless steel
  • Non-magnetic
  • Microfine, pointed tips
Specifications
Specifications
Strong, Fine
Stainless Steel
Forceps, Swiss Micro Jeweler
1
4.75 in.
120 mm
Each