Integra™ Miltex™ Swiss Jeweler-Style Forceps
Supplier:  Integra™ 17307X
 View more versions of this product
Catalog No. 12-460-250
Explore available promotions
Description
Description
  • Stainless steel
  • Non-magnetic
  • Microfine, pointed tips
Specifications
Specifications
Micro Fine, Curved
Stainless Steel
Forceps, Swiss Micro Jeweler
7
4.5 in.
114 mm
Each