missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Interference Check Standard 18 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC INTER18100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
K | 20,000μg/mL |
As | 1000μg/mL |
Pb | 1000μg/mL |
Tl | 1000μg/mL |
Se | 500μg/mL |
Ag | 300μg/mL |
Ba | 300μg/mL |
Cd | 300μg/mL |
Co | 300μg/mL |
Cr | 300μg/mL |
Cu | 300μg/mL |
Ni | 300μg/mL |
V | 300μg/mL |
Zn | 300μg/mL |
Mn | 200μg/mL |
Be | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Interference Check Standard 18 | |
125 mL | |
Certified Reference Material | |
5% HNO3 | |
Environmental |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
AA, ICP | |
Various |
Product intended for laboratory use only