Intertek™ MIC Test Kit, Petroleum Microbiology Media: TGB DFS Item
Thioglycollate Medium, for anaerobic and facultative anaerobic heterotrophic bacteria
Supplier:  Intertek™ MK120C
 View more versions of this product
Catalog No. 08629006C
Explore available promotions
Description
Description
For use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
Microbiology Media
Vial
United States
Cultivation of heterotrophic anaerobic bacteria, MIC testing
12 Vials