missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ 1 Kb Plus DNA Ladder DFS Item

Powerful tool for estimating molecular weight of linear, double-stranded DNA fragments

$269.00 - $818.00

Specifications

Concentration 0.5 μg/μL
Gel Type Agarose
Product Type DNA Ladder
Ready to Load No
Sample Loading Volume 1 mL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10787018 DFS Item
View Documents
Invitrogen™
10787018
1 μL DNA Ladder (500 ng), 1 μL Sample Loading Buffer, 500 Applications
Each for $269.00
 
10787026 DFS Item
View Documents
Invitrogen™
10787026
4 x 500 μL DNA Ladder, 4 x 1 mL Sample Loading Buffer, 2000 Applications
Each for $818.00
 
Description

Description

One Kb Plus DNA ladder is composed of 20 highly purified, double-stranded DNA bands spanning 100bp to 12,000bp. This ladder has 12 evenly spaced bands ranging from 1Kb to 12Kb, quick orientation band at 1,650bp that forms distinct doublet with 2Kb band, and seven bands of round sizes below 1Kb. One Kb Plus DNA ladder is compatible with agarose gels cast in lab as well as E-Gel™ pre-cast agarose gels. One Kb Plus DNA ladder is most popular DNA ladder used to estimate molecular mass of double-stranded DNA bands from PCR, restriction digests, and other molecular biology protocols. Unlike some other DNA ladders, 1Kb Plus DNA ladder has been specially designed with bands of round, easy-to-remember sizes. Logical band layout, and quick identification doublet, allow to estimate size of band at glance.

  • Round, easy-to-remember band sizes
  • Broad size range (100bp–12Kb)
  • Compatible with all agarose gels
  • Less waste, sustainable packaging

  • Popular and easiest-to-use ladder
  • Single ladder for most of needs
  • Simplified format for use with all DNA agarose gels
  • DNA stain and radiolabel compatible

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

0.5 μg/μL
DNA Ladder
1 mL
500 μL
Sustainable packaging
Individual chromatography-purified DNA fragments
Agarose
No
100 to 15000 bp
Agarose gel
Approved for shipment at Room Temperature or on Dry Ice
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.