Learn More
Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity
Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates.
Supplier: Invitrogen™ 12346094
Description
Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates. It includes an enzyme blend of Platinum Taq DNA Polymerase and a proofreading enzyme, plus AccuPrime accessory proteins for improved PCR fidelity, yield, and specificity over other hot-start DNA polymerases.
Features of AccuPrime Taq DNA Polymerase, High Fidelity
- High specificity and yield for robust PCR amplification
- 9-fold higher fidelity than Taq DNA polymerase alone
- Mimimal optimization steps, even with non-optimized primer sets
- Efficient amplification of targets over a broad size range up to 20 kb
How it works
Pyrococcus species GB-D polymerase is a proofreading enzyme that possesses a 3' to 5' exonuclease activity. Mixing this enzyme with Taq DNA Polymerase increases fidelity and allows amplification of simple and complex DNA templates over a large range of target sizes.
The Platinum antibody complexes with Taq DNA polymerase and inhibits activity at room temperature. Activity is restored after the initial denaturation step, providing an automatic hot start PCR.
The thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, preventing mispriming and enhancing PCR specificity and yield.
Specifications
• AccuPrime Taq DNA Polymerase, High Fidelity (1 x 200 μL at 5 U/μL) • 10X AccuPrime PCR Buffer I (4 x 1.25 mL) • 10X AccuPrime PCR Buffer II (4 x 1.25 mL) • 50 mM MgSO4 (4 x 1.25 mL) Store at -10°C to -30°C. |
|
Low | |
Standard | |
Dry Ice | |
9X | |
1000 Reactions | |
Separate Components |
Primer-probe | |
AccuPrime Taq DNA Polymerase High Fidelity | |
DNA Polymerase | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
Mixed, 3'-A | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.