missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIAP (Calf Intestinal Alkaline Phosphatase), 1 U/μL
Phosphomonoesterase that removes 3' and 5' phosphates from DNA and RNA
Supplier: Invitrogen™ 18009027
Description
- Concentration: 20 units/μL
- Source: Purified from calf intestinal mucosa
- Performance and Quality Testing: Endodeoxyribonuclease, 3' exodeoxyribonuclease, and ribonuclease assays; dephosphorylation efficiency measured in a transformation assay
- Unit Definition: One unit hydrolyzes 1μmol of 4-nitrophenyl phosphate in 1 min. at 37°C
- Unit Reaction Conditions: 1M diethanolamine buffer, 10mM 4-nitrophenyl phosphate, 0.25mM MgCl2 (pH 9.8) in 900μL for 10 min. at 37°C
Dephosphorylation of 5'-phosphorylated termini of vector DNA to prevent self-ligation; Dephosphorylation of 5' termini of nucleic acids prior to forward reaction with kinase; Cloning; Restriction Enzyme Cloning
Specifications
| 1 U/μL | |
| Approved for shipment on Wet or Dry Ice | |
| Diethanolamine Buffer | |
| Alkaline Phosphatase |
| Calf Intestinal Alkaline Phosphatase (1 unit/μL) is supplied with a vial of 10X dephosphorylation buffer [500 mM Tris-HCl (pH 8.5), 1 mM EDTA], vial of dilution buffer [50% glycerol, 25 mM Tris-HCl (pH 7.6), 1 mM MgCl2 , and 0.1 mM ZnCl2 ]. Store Calf Intestinal Alkaline Phosphatase at -20°C. | |
| CIP | |
| 1000 U |
For Research Use Only. Not for use in diagnostic procedures.