missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DNA Polymerase I DFS Item

DNA polymerase with 5'→3' and 3'→5' exodeoxyribonuclease activities

Supplier:  Invitrogen™ 18010025

 View more versions of this product

Catalog No. 18010025


Explore available promotions

Description

Description

Performance and quality testing
Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50mM potassium phosphate (pH 7.0), 6.7mM MgCl2 , 1mM 2-mercaptoethanol, 80μg/mL template/primer, 32μM dTTP, 69nM [3H]dTTP, and enzyme in 100μL for 30 min at 37°C. Also incorporates biotinylated nucleotides.

  • Applications: DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5' overhangs
  • Source: purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Cloning, Restriction Enzyme Cloning

Specifications

Specifications

1000 units of DNA polymerase in a storage buffer of 50 mM potassium phosphate (pH 7.0), 100 mM KCl, 1 mM DTT, and 50% (v/v) glycerol. It is supplied at 5–10 units/μl.

Store at -5°C to -30°C.
DNA Polymerase I
1000 U
No
DNA Polymerase
3' - 5', 5' - 3'
Wet Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.