missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment DFS Item

DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities

Supplier:  Invitrogen™ 18012021

 View more versions of this product

Catalog No. 18012021


Explore available promotions

Description

Description

Applications:
Fill-in of 5' overhangs . Synthesis of probes by random primers labeling method. Sequencing single- and double-stranded DNA. Site-directed mutagenesis.

Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.

Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.

Unit Definition:
One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template•primer.

Array CGH, Cloning, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Restriction Enzyme Cloning

Specifications

Specifications

DNA Polymerase I, Large (Klenow) Fragment is supplied with a vial of 10X REact™ 2 Buffer [500 mM Tris-HCl (pH 8.0), 100 mM
MgCl2 , 500 mM NaCl], vial of dilution buffer. Store at -20°C.
DNA Polymerase I
100 Units
No
DNA Polymerase
3' to 5'
Approved for shipment on Wet or Dry Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.