Invitrogen™ DNaseAlert™ QC System
Provides sensitive detection of DNases in a simple, easy-to-use fluorimetric assay with real time results
Supplier: Invitrogen™ AM1970
Specifications
Lyophilized | |
Dry Ice | |
DNase Control Reagent |
Store DNaseAlert™,10X NucleaseAlert Buffer, DNase I and TE Buffer pH 7.0 at -20°C. |
|
480 assays | |
Ambion™, RNaseAlert™ |
For Research Use Only. Not for use in diagnostic procedures.