missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Dynabeads™ CD3 DFS Item

Dynabeads™ CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, mononuclear cells (MNC), and tissue digests.

Supplier:  Invitrogen™ 11151D

Catalog No. 11151D


Explore available promotions

Includes: 5mL Dynabeads CD3 coated with anti-CD3 monoclonal antibody.

Description

Description

Dynabeads™ CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, mononuclear cells (MNC), and tissue digests. Advantages of Dynabeads™ CD3:

• Fast and efficient isolation of CD3+ T cells from any sample—no columns required
• High purity, yield, and viability of positively isolated CD3+ T cells for molecular downstream assays
• Fast and efficient depletion of CD3+ T cells from any sample

About Dynabeads™ CD3
Dynabeads™ CD3 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD3 membrane antigen mainly expressed mainly on human mature T cells. Because of the bead size, Dynabeads™ CD3 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads™ CD3 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD3+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human T cell product offerings.

Magnetic bead–based separation offers easy handling
Dynabeads™ CD3 is added to the sample under continuous mixing to optimize the binding of the Dynabeads™ to the target cells. By placing the sample on a magnet, it separates the bead-bound target cells from the rest of the sample in just 1–2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2–3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

T Cells (Whole Population)
Antibody Coated Bead
Processes ∼2 x 109 cells total
DYNAL™, Dynabeads™
PBMC, Blood
1 x 107 PBMCs per isolation
4.5 μm
This Product Contains: 5 mL Dynabeads™ CD3 coated with anti-CD3 monoclonal antibody.

Storage Conditions: 2°C to 8°C. Do not freeze.
Depletion, positive isolation for molecular applications
>95%
Research Grade
Room Temperature
Human
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.