missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Electroporation Cuvettes, 0.4 cm DFS Item

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are specifically designed help achieve high DNA transformation efficiency during the introduction of DNA into bacterial, yeast, fungal, and mammalian cells.

Supplier:  Invitrogen™ P46050

Catalog No. P46050


Explore available promotions

Description

Description

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are specifically designed help achieve high DNA transformation efficiency during the introduction of DNA into bacterial, yeast, fungal, and mammalian cells.

Our electroporation cuvettes are precision manufactured from the highest quality materials to ensure optimal and reproducible transformation results.

Electroporation cuvette features
• Compatible with virtually all common electroporation devices
• Gamma-irradiated and individually packaged to ensure sterility
• Color-coded snap cap for easy identification of gap size (red cap for 0.4 cm cuvette)

Note: Electroporation cuvettes are recommended for use with all of our electroporation competent cells.

Specifications

Specifications

Electroporation Device
0.4 cm
• 50 individually wrapped, sterile cuvettes

Store at room temperature.

Electroporation Cuvette
50 Cuvettes/Bag
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.