missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Taq DNA Polymerase, recombinant DFS Item

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer.

Supplier:  Invitrogen™ 10342046

 View more versions of this product

Catalog No. 10342046


Explore available promotions

Description

Description

Taq DNA Polymerase, recombinant, is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. This recombinant enzyme is purified from the cloned Thermus aquaticus DNA polymerase gene expressed in E. coli and consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5' to 3' DNA polymerase activity and a 5' to 3' exonuclease activity.

Features

  • A choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • Enzyme is licensed and qualified for PCR

Applications

  • Amplification of DNA from complex genomic, viral, and plasmid templates
  • RT-PCR
  • Sequencing ssDNA
  • Cycle sequencing

Notes

  • One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of deoxyribonucleotide into DNA in 30 minutes at 74°C.
  • For superior PCR performance, DreamTaq DNA Polymerase is recommended.
Specifications

Specifications

Taq DNA Polymerase (5 U/μL), 3 x 100 μL
• 10X PCR buffer, 7.5 mL
• 50 mM MgCl2, 3 mL

Store at -10°C to -30°C in a non-frost-free freezer.
Guaranteed stable for 6 months when stored properly.
Low
Taq DNA Polymerase
5' - 3'
3 x 500 Units
Standard PCR
1X
1500 Reactions
Separate Components
DNA
Stand-alone enzyme
qPCR, Standard PCR
Standard
DNA Polymerase
Dry Ice
5 U/μL
No
3'-A
5 kb or less
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.