Invitrogen™ Lipofectamine™ 2000 Transfection Reagent DFS Item
Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines
Supplier:  Invitrogen™ 11668027
 View more versions of this product
Catalog No. 11668027
Explore available promotions
Description
Description
Lipofectamine™ 2000 Transfection Reagent is a versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines. Researchers use Lipofectamine™ 2000 Reagent for siRNA- and shRNA-based gene knockdown experiments, as well as for gene expression studies. Lipofectamine™ 2000 Transfection Reagent is our best transfection reagent available for the co-transfection of siRNA and plasmid DNA.
With Lipofectamine™ 2000 Transfection Reagent, you'll get:
  • Exceptional transfection efficiency in the broadest range of cell lines and the highest levels of recombinant protein expression (see table)
  • Superior performance for co-transfection of siRNA and plasmid DNA
  • Proven efficacy in the presence of serum—eliminates the need to change media following transfection
  • Reliable performance for high-throughput applications
  • The best choice for establishing stable cell lines
    A high-performance transfection reagent for gene expression and gene silencing
    Lipofectamine™ 2000 Transfection Reagent works effectively with all common cell lines as well as with many challenging ones, and can be used in media with or without serum. For gene silencing, Lipofectamine™ 2000 Transfection Reagent's high-efficiency transfections provide the high levels of gene knockdown needed to achieve convincing results. Lipofectamine™ 2000 Transfection Reagent is the number one choice for co-transfection, given its effectiveness for transfecting both siRNA and plasmid DNA. Lipofectamine™ 2000 Transfection Reagent is easy to use—simply mix with nucleic acid and add to cell culture.
    Ideal for high-throughput work
    Simplicity and speed combined with high transfection efficiency make Lipofectamine™ 2000 Transfection Reagent ideal for transient protein expression or high-throughput RNAi transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
  • Specifications
    Specifications
    Contains one vial (0.75 ml) Lipofectamine™ 2000 Reagent. Store at 2–8°C. Do not freeze.
    6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
    Lipid-based Transfection
    Sustainable packaging
    Lipofectamine™
    0.75 mL
    Approved for shipment at Room Temperature or on Wet Ice
    Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
    Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
    Transfection
    High-throughput Compatible
    Transfection Reagent
    Yes
    Videos
    SDS
    Documents
    Documents
    Promotions
    Promotions
    For Research Use Only. Not for use in diagnostic procedures.