missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PCR SuperMix DFS Item

PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

Supplier:  Invitrogen™ 10572014

Catalog No. 10572014


Explore available promotions

Description

Description

PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Simply add template and primers, reducing set-up time by half. PCR SuperMix contains Mg2+, dNTPs, and recombinant Taq DNA Polymerase at concentrations sufficient for routine PCR of fragments up to 5 kb. PCR SuperMix is supplied at a 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions.

For superior PCR performance, we recommend DreamTaq DNA Polymerase.

Specifications

Specifications

• 4 vials of PCR SuperMix, 1.125 mL each

Store at -20°C.
Low
Standard
PCR SuperMix
Standard PCR
1X
100 Reactions
SuperMix or Master Mix
DNA
Tube
Taq DNA Polymerase
5' - 3'
Wet or Dry Ice
1.1X
No
3'-A
5 kb or less
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.