missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ PCR SuperMix High Fidelity DFS Item

Platinum PCR SuperMix, High Fidelity, is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.

Supplier:  Invitrogen™ 12532016

 View more versions of this product

Catalog No. 12532016


Explore available promotions

Description

Description

Platinum PCR SuperMix, High Fidelity, is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Designed for high-specificity, high-fidelity DNA amplification, Platinum PCR SuperMix reduces reaction set-up time and only requires the addition of template and primer.

High fidelity is provided by a mixture of Taq DNA polymerase and the proofreading Pyrococcus species GB-D polymerase. High specificity is achieved by anti-Taq antibodies, allowing for automatic hot-start PCR.

Benefits of Platinum PCR SuperMix, High Fidelity

  • Six-times higher fidelity than Taq DNA polymerase
  • Amplifies fragments up to 15 kb
  • Minimizes PCR optimization
  • Room-temperature reaction assembly

Applications

  • High-fidelity PCR
  • Cloning
  • Mutagenesis

Notes

  • Platinum PCR SuperMix, High Fidelity, produces products that are mixed blunt and 3-A' ends; however, the majority of ends will have a 3-A' overhangs.
  • Hot-start capability increases specificity and yield and allows for room-temperature reaction assembly.
Specifications

Specifications

Platinum PCR SuperMix High Fidelity (4 x 1.125 mL)

Store at -20°C in a non-frost-free freezer.
Platinum Taq DNA Polymerase High Fidelity
100 rxns
Hot-start PCR, High-fidelity PCR
6X
100 Reactions
SuperMix or Master Mix
Low
Standard
Wet or Dry Ice
1.1X
Built-In Hot Start
Mixed
15 kb or less
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.