Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit DFS Item
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Supplier:  Invitrogen™ 11732020
 View more versions of this product
Catalog No. 11732020
Explore available promotions
Description
Description
SuperScript™ III Platinum™ One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR™ Green I detection.
  • The SuperScript III Platinum One-Step qRT-PCR Kit’s proven formula delivers reliable, accurate target detection when it matters most
  • A convenient, one-step format and consistent performance make it ideal for a number of gene expression applications, including infectious disease surveillance
  • The kit has been validated by several global public health organizations and has long been the choice of the US CDC for infectious disease surveillance
  • SuperScript III RT for higher temperature cDNA synthesis when addressing difficult RNA secondary structure
  • Platinum Taq DNA Polymerase with hot-start technology for improved specificity
  • LUX fluorogenic primers and dual-labeled fluorogenic probes for effective detection performance
  • Validated performance on multiple real-time instrument platforms, including rotor-based systems
  • Multiple SuperScript III Platinum One-Step qRT-PCR Kit configurations are available
  • Store components at -20°C, ROX Reference Dye must be stored in the dark
Specifications
Specifications
• 100 μl SuperScript™ III/Platinum™ Taq Mix
• 2 × 1.25 ml 2X Reaction Mix
• 1 ml Magnesium Sulfate (50 mM)
• 100 μl ROX Reference Dye (25 μM)
Store components at -20°C. ROX Reference Dye must be stored in the dark.
1-step RT-qPCR
Standard
Platinum™, SuperScript™
100 reactions
100 Reactions
100 Reactions
Primer-probe
Taq DNA Polymerase
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
One-Step qRT-PCR Kit
Dry Ice
Gene Expression, Pathogen Detection, Virus Detection
RNA
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.