missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PLUS™ Reagent
Used in conjunction with various transfection reagents, such as Lipofectamine™ Reagent, to improve transfection efficiency in adherent cell lines
Supplier: Invitrogen™ 11514015
Includes: Contains one vial (0.85mL) PLUS Reagent.
Description
- Some customers find that when using PLUS Reagent, less optimization of DNA concentration is needed to achieve high transfection performance
- Addition of PLUS Reagent may also reduce quantities of DNA and lipid required by approximately one-half compared to lipid alone
- PLUS Reagent was specifically designed to expand array of transfectable cell types
- When used with Lipofectamine Reagent, PLUS Reagent has been demonstrated to improve the transfection of BHK-21, NIH 3T3, and other cell lines
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet Ice
Specifications
Contains one vial (0.85 ml) PLUS™ Reagent. Store at 4°C. Do not freeze. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
Not High-throughput Compatible (Manual) | |
Reagent | |
Yes |
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
Plasmid DNA, RNAi Plasmids (shRNA, miR) | |
Transfection | |
PLUS™ | |
0.85 mL | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.