Invitrogen™ PureLink™ Genomic DNA Mini Kit DFS Item
The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format.
Supplier:  Invitrogen™ K182000
 View more versions of this product
Catalog No. K182000
Explore available promotions
Description
Description
The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink™ Genomic DNA Mini Kit include:
Kit flexibility—one kit works with a variety of sample types and sizes
Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
Improved design—optimized spin-column design and buffer formulation for better yield and purity
Multiple sample sources, one kit
The PureLink™ Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.
Optimized components and protocols
The PureLink™ Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use. Columns are newly configured to bind and release DNA more efficiently. Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.
Higher yields and purity
With PureLink™ Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells. The enhanced PureLink™ Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ∼1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine.
Specifications
Specifications
• 2.0 mL PureLink™ Genomic Lysis/Binding Buffer
• 1.8 mL PureLink™ Genomic Digestion Buffer
• 2.0 mL PureLink™ Genomic Wash Buffer 1
• 1.5 mL PureLink™ Genomic Wash Buffer 2
• 2.0 mL PureLink™ Genomic Elution Buffer
• 0.4 mL RNase A (20 mg/mL)
• 0.2 mL Proteinase K (20 mg/mL)
• 10 PureLink™ Spin Columns with Collection Tubes
• 20 PureLink™ Collecion Tubes (2.0 mL)
Store all components at room temperature.
Mini
Genotyping, Real-Time Quantitative PCR (qPCR), Southern Blotting, Sequencing, PCR
100 μL to 1 mL Blood Samples (Medium-Scale)
Genomic DNA Mini Kit
Room Temperature
Silica Spin Column
Bacteria, Blood, Cells, Tissue
Not High-throughput Compatible (Manual)
PureLink™
10 Preps
Genomic DNA
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.