missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ HiPure Plasmid Maxiprep Kit DFS Item

The PureLink HiPure Plasmid Maxiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli.

Supplier:  Invitrogen™ K210006

 View more versions of this product

Catalog No. K210006


Explore available promotions

Description

Description

The PureLink HiPure Plasmid Maxiprep Kit is designed to isolate transfection-grade plasmid DNA from E. coli. The Maxiprep Kit protocol will typically yield 500–850 μg plasmid DNA from 100–200 mL bacterial culture, at purity that is comparable to that achieved by two passes through cesium chloride gradients.

  • Labor and time saving- no need for extra steps to remove endotoxin and other contaminants
  • High-purity, low-endotoxin product- pure enough for mammalian cell transfection
  • Versatility—purify any type and size of plasmid DNA, including BAC, bacmids, and ssM13 DNAs

Anion-exchange chromatography for plasmid purification

  • PureLink HiPure Plasmid DNA Purification Kits use patented anion-exchange resin to purify plasmid DNA to a level equivalent to two passes through CsCl gradients
  • Resin combines excellent capacity with a fast flow rate, high resolution, high yield, and efficient endotoxin removal
  • Plasmid preparations can typically be completed in less than 2 hours

Contaminant-free DNA

  • Unlike cesium chloride gradient protocols, PureLink HiPure Plasmid Purification Systems do not use troublesome solvents to work with and to dispose of
  • Typically, plasmid DNA prepared using the PureLink HiPure Plasmid Midiprep Kit will have an A260/A280 ratio of >1.80, indicating that the DNA is reasonably free of proteins that might interfere with downstream applications
  • Endotoxin levels are typically within 0.1–1 EU/μg, making this plasmid DNA ideal for mammalian cell transfection
  • Purification time is two hours
Specifications

Specifications

• 100 mL Resuspension Buffer (R3)
• 550 μL RNase A
• 100 mL Lysis Buffer (L7)
• 100 mL Precipitation Buffer (N3)
• 300 mL Equilibration Buffer (EQ1)
• 2 × 300 mL Wash Buffer (W8)
• 200 mL Elution Buffer (E4)
• 30 mL TE Buffer
• 10 HiPure Columns
• 3 Column Holders

Store all components at room temperature.
Anion Exchange Resin
Bacterial Culture
BAC DNA, Plasmid DNA
Not High-throughput Compatible (Manual)
<40kb, Low Copy Plasmid, High Copy Plasmid, BAC
PureLink™
10 Preps
Room Temperature
PureLink™
1000 μg
Column
Maxi
15 mL
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
10 Preps
> 200 μg (Large-Scale) Plasmid DNA
Plasmid MaxiPrep Kit
Transfection Grade
BAC DNA, Plasmid DNA
2 hr.
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.