Invitrogen™ PureLink™ HQ Mini Plasmid DNA Purification Kit DFS Item
The PureLink HQ Mini Plasmid Purification Kit is designed for the isolation of high-quality plasmid DNA that is suitable for restriction enzyme digestion, PCR, sequencing, bacterial cell transformation, and mammalian cell transfection.
Supplier:  Invitrogen™ K210001
Catalog No. K210001
Explore available promotions
Includes: Sufficient PureLink columns, collection tubes, elution tubes, and reagents for 100 purifications.
Description
Description
The PureLink HQ Mini Plasmid Purification Kit is designed for the isolation of high-quality plasmid DNA that is suitable for restriction enzyme digestion, PCR, sequencing, bacterial cell transformation, and mammalian cell transfection. Using the kit, plasmid DNA can be isolated from varying amounts of bacterial cells.
The PureLink HQ Mini Plasmid Purification Kit has the following advantages:
• Higher spin column capacity for plasmid DNA as compared to other commercially available plasmid purification systems
• Designed to isolate high-quality plasmid DNA in less than an hour
• Minimal genomic DNA contamination of the purified sample
• Reliable performance of the purified DNA in downstream applications
Process overview
Bacterial cells are pelleted, resuspended, and lysed. The lysate is then neutralized and conditions are adjusted for subsequent binding. After clarification by centrifugation, the lysate is processed through the PureLink spin column. The DNA binds to the silica-based membrane in the column, and impurities are removed by a single wash step. The DNA is then eluted in Elution Buffer or water.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Spin Column
Spin Column
Mini
Plasmid DNA
Not High-throughput Compatible (Manual)
< 100 μg (Small-Scale) Plasmid DNA
Plasmid DNA Purification Kit
Room Temperature
The PureLink HQ Mini Plasmid DNA Purification Kit includes sufficient PureLink columns, collection tubes, elution tubes, and reagents for 100 purifications. Store components at room temperature. Store Resuspension Buffer after addition of RNase A at +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Silica Spin Column
Bacterial Culture
PCR, Transfection, Sequencing, Transformation, Cloning
100 Preps
PureLink™
100 Preps
Plasmid DNA
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.