missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ Pro 96 Genomic DNA Purification Kit DFS Item

The PureLink™ Pro 96 Genomic DNA Purification Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.

Supplier:  Invitrogen™ K182104A

Catalog No. K182104A

Explore available promotions

Includes:

80mL PureLink™ Pro 96 Genomic Lysis/Binding Buffer, 70mL PureLink™ Pro 96 Genomic Digestion Buffer, 2 x 100mL PureLink™ Pro 96 Genomic Wash Buffer 1, 2 x 75mL PureLink™ Pro 96 Genomic Wash Buffer 2, 160mL PureLink™ Pro 96 Genomic Elution Buffer, 8mL RNase A (20mg/mL), 8mL PureLink™ Pro 96 Proteinase K (20mg/mL), 4 PureLink™ Genomic DNA Filter Plate, 2 x 6 96 Deep Well Blocks, 20/pack PureLink™ Foil Tape


Description

Description

Kit flexibility: One kit works with a variety of sample types and sizes
Ultraclean gDNA: Excellent A260/A280 and A260/A230 absorbance ratios, regardless of sample source
Improved design: Optimized, semi-skirted, 96-well plates compatible with centrifugation and most benchtop and robotic vacuum manifolds

Multiple sample sources, one kit
The PureLink™ Pro 96 Genomic DNA Purification Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual. Examples of sample sources and amounts that can be used with the PureLink™ Pro Genomic DNA Purification Kit include mammalian cells and tissue, FFPE tissue, Oragene™-preserved saliva, mouse and rat tails, non-nucleated whole blood (human, mouse), nucleated whole blood (bird), blood spot on paper, bacterial cells, and yeast cells.

Improved plate design
The PureLink™ Pro 96 Genomic DNA Purification Kit provides optimized, semi-skirted, 96-well plates, for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. Sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation (at ∽2 x g), or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.

Higher yields and purity
The PureLink™ Pro 96 Genomic DNA Purification Kit outperforms competitor products in both DNA yield and purity for multiple sample types, including blood. You are also able to get greater yields of higher-purity gDNA (determined by A260/A280 measurements) no matter what your sample type is; from bacteria to tissues to blood to cells. The enhanced PureLink™ Lysis and Wash Buffers help consistently deliver purified gDNA samples with absorbance reading ratios of ≅1.9 for both A260/A280 and A260/A230, indicating no contamination carryover of protein or guanidine.

Not High Throughput-Compatible (Manual), High Throughput-Compatible

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

• 80 mL PureLink Pro 96 Genomic Lysis/Binding Buffer; room temperature
• 70 mL PureLink Pro 96 Genomic Digestion Buffer; room temperature
• 2 x 100 mL PureLink Pro 96 Genomic Wash Buffer 1; room temperature
• 2 x 75 mL PureLink Pro 96 Genomic Wash Buffer 2; room temperature
• 160 mL PureLink Pro 96 Genomic Elution Buffer; room temperature
• 8 mL RNase A; room temperature
• 8 mL PureLink Pro 96 Proteinase K; room temperature, or 4°C for long-term storage
• 4 PureLink Genomic DNA Filter Plates; room temperature
• 2 x 6 plates 96 Deep Well Blocks; room temperature
• 20/pack PureLink Foil Tape; room temperature

Mini
50 to 200 μL
Biomek™ FX, Eppendorf epMotion™, TECAN Freedom Evo™, TECAN Genesis™, Robotic Liquid Handling Systems
High-throughput Compatible
Room Temperature
0.5 mg (Binding capacity)
Silica Plate
Bacteria, Cells, Dried Blood Spots, FFPE (Formalin-fixed paraffin-embedded), Mouse/Rat Tail, Saliva, Buccal Swab, Tissue, Whole Blood, Yeast
gDNA
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
4 x 96 Preps
45 min. (Including sample homogenization)
Bacteria: ≤109 cells
Buccal Swab: 1
Cells: ≤106
Dried Blood Spots: ≤5 punches
FFPE: ≤8 sections of 5-15 μM thickness
Mouse Tail: 1 cm
Rat Tail: 0.5 cm
Saliva (Oragene™ preserved): ≤200 μL
Tissue: ≤25 mg
Whole Blood: ≤200 μL
Yeast: ≤107 cells
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.