missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Qubit™ Assay Tubes DFS Item

For use with the Qubit Fluorometer

Supplier:  Invitrogen™ Q32856

Catalog No. Q32856


Explore available promotions

Description

Description

  • Qubit assay tubes are 500μL, thin-walled polypropylene tubes for use with the Qubit Fluorometer
  • 500 tubes per package

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store at room temperature.
Assay Tube
500 tubes
Room Temperature
Qubit™
Qubit Fluorometer
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.