missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Ribonuclease H DFS Item

Specifically degrades RNA strand of RNA-DNA hybrid to produce 5' phosphate-terminated oligoribonucleotides and single-stranded DNA

Supplier:  Invitrogen™ 18021014

 View more versions of this product

Catalog No. 18021014

Explore available promotions

Description

Description

  • Source: Purified from E. coli expressing the E. coli RNase H gene on a plasmid
  • Performance and quality testing: Ribonuclease, nonspecific endodeoxyribonuclease, 3' and 5' exodeoxyribonuclease
  • Unit definition: One unit hydrolyzes 1nmol of RNA in 3H-labeled poly(A)poly(dT) to acid-soluble material in 20 min. at 37°C
  • Unit reaction conditions: 20mM Tris-HCl (pH 7.5), 0.1M KCl, 10mM MgCl2 , 0.1mM DTT, 5% (w/v) sucrose, 0.5nmol 3H-labeled poly(A)poly(dT), and enzyme in 50μL for 20 min. at 37°C

Removal of mRNA during second-strand cDNA synthesis; Removal of poly(A) sequences from mRNA in the presence of oligo(dT); Oligodeoxyribonucleotide-directed cleavage of RNA; PCR & Real-Time PCR; Reverse Transcription

Specifications

Specifications

2 U/μL
Approved for shipment on Wet or Dry Ice
30 U
1 vial of Ribonuclease H containing 30 units supplied at a concentration of 2U/μl

Store at -20°C in a non-frost-free freezer.
RNase
Ribonuclease H
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.