missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO DFS Item

Extremely sensitive stain for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels

Supplier:  Invitrogen™ S7567

 View more versions of this product

Catalog No. S7567


Explore available promotions

Description

Description

SYBR Green I offers greater sensitivity for dsDNA, making it especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR Green I stain is ideal for use with laser scanners.
Product use 1mL stains 100 minigels.

  • Sensitivity: at least four times greater than ethidium bromide for DNA in agarose gels
  • Ease of use: gels soaked in diluted stain can be visualized without desalting
  • Compatibility: with UV transilluminators, gel documentation systems, and laser scanners
  • Flexibility: for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
Nucleic Acid Gel Stain
Room Temperature
In-Gel Detection
SYBR Green I
1 mL
DNA
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.