Learn More
Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE
Developed specifically for reduced mutagenicity to be less hazardous. Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE is safer and more sensitive than ethidium bromide for staining DNA in agarose or acrylamide gels. The premixed solution is available in 1L and 4L units.
$201.00 - $622.00
Specifications
Content And Storage | • Supplied as a ready-to-use solution in 1X TAE Store at room temperature in the original container. |
---|---|
Detection Location | In-Gel Detection |
Detection Method | Fluorescence |
Label or Dye | SYBR Safe |
Product Type | DNA Gel Stain |
Description
SYBR Safe DNA gel stain was developed specifically for reduced mutagenicity—to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels. SYBR Safe stain is not only less mutagenic than ethidium bromide, but SYBR Safe stain's detection sensitivity is better than that of ethidium bromide. SYBR Safe stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∽280 and ∽502nm, and an emission maximum at ∽530nm.
- Less hazardous
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room temperature
Specifications
• Supplied as a ready-to-use solution in 1X TAE Store at room temperature in the original container. |
|
Fluorescence | |
DNA Gel Stain | |
Room Temperature |
In-Gel Detection | |
SYBR Safe | |
Less hazardous | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.