missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector, without competent cells DFS Item

Provides quick, one-step cloning strategy for directly inserting Taq-amplified PCR product into plasmid vector

Supplier:  Invitrogen™ K202040

 View more versions of this product

Catalog No. K202040


Explore available promotions

Description

Description

The TA Cloning Kit uses the pCR2.1 cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast and convenient: 15-minute, room-temperature ligation
  • Efficient: blue/white screening and >80% clones with correct insert
  • Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free: eliminates any enzymatic modifications of the PCR product
  • Streamlined: does not require the use of PCR primers that contain restriction sites

  • Provides 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 promoter for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of insert
  • M13 forward and reverse primer sites for sequencing
  • How TA Cloning Works
  • Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to 3' ends of PCR products
  • Linearized vector supplied in this kit has single 3' deoxythymidine (T) residues
  • Allows PCR inserts to ligate efficiently with vector

Cloning, PCR Cloning

Specifications

Specifications

Not Included
TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls.

Store all components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
PCR Cloning
TA Cloning™
T7
pCR2.1
TA Cloning
Kit
40 Reactions
Cloning Kit
40 rxns
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.